Zink 99,99 Metals basis, sputtering target, Ø 50.8 mm (2.0 in), Dicke 3.18 mm (0.125 in)

Lieferant: Thermo Fisher Scientific

41655.KS
41655.KSEA 282 EUR
41655.KS
Zink 99,99 Metals basis, sputtering target, Ø 50.8 mm (2.0 in), Dicke 3.18 mm (0.125 in)
Zinc

Purity: 99.99% CAS No.: 7440-66-6 Odour: Odourless MDL No.: MFCD00011291


Magnetron sputtering source

Formel: Zn
Molecular Weight: 65.39 g/mol
Siedepunkt: 908
Schmelzpunkt: 420
Dichte: 7.14 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00011291
CAS-Nummer: 7440-66-6
EINECS: 231-175-3
Merck Index: 12,10255

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR