Zink 99,99 Metals basis, sputtering target, Ø 50.8 mm (2.0 in), Dicke 6.35 mm (0.250 in)
Lieferant: Thermo Fisher Scientific
41656.KS
41656.KSEA
456
EUR
41656.KS
Zink 99,99 Metals basis, sputtering target, Ø 50.8 mm (2.0 in), Dicke 6.35 mm (0.250 in)
Zinc
Purity: 99.99%
CAS No.: 7440-66-6
Odour: Odourless
MDL No.: MFCD00011291
Formel:
Zn Molecular Weight: 65.39 g/mol Siedepunkt: 908 Schmelzpunkt: 420 Dichte: 7.14 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011291 CAS-Nummer: 7440-66-6 EINECS: 231-175-3 Merck Index: 12,10255 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...