Bismuth titanium oxide ≥99,9% (Metall-Basis)
Lieferant: Thermo Fisher Scientific
41669.18
41669.30
41669.18EA
98.6
EUR
41669.18
41669.30
Bismuth titanium oxide ≥99,9% (Metall-Basis)
Bismuth titanium oxide
Purity: 99.9%
CAS No.: 12233-34-0
Molecular Formula: Bi2Ti4O11
Formula Weight: 785.42
Physical Form: -325 Mesh Powder
Odour: Odourless
MDL No.: MFCD01310393
Formel:
Bi₁₂TiO₂₀ Lagertemperatur: Raumtemperatur |
MDL:
MFCD01310393 CAS-Nummer: 12233-34-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...