Ethylmagnesiumbromid 3 M in Äther
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
verpackt unter Argon in wiederversiegelbaren ChemSeal™ Flaschen
CAS No.: 925-90-6
Molecular Formula: C2H5BrMg
Formula Weight: 133.28
Physical Form: Liquid
Density: 1.020
Sensitivity: Air and Moisture Sensitive
Appearance: Pale yellow to brown
Odour: Ether-like
Melting Point: -116.3°
Boiling Point: 34.6°
UN No.: 3399
MDL No.: MFCD00000043
Formel:
C₂H₅BrMg Siedepunkt: 34,6 °C Dichte: 1,020 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00000043 CAS-Nummer: 925-90-6 EINECS: 213-127-3 UN: 3399 ADR: 4.3,I |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...