Kupfer(II)oxid ≥99,7% (Metall-Basis)

Lieferant: Thermo Fisher Scientific
Achtung

41692.22 41692.36 41692.A3
41692.22EA 75.8 EUR
41692.22 41692.36 41692.A3
Kupfer(II)oxid ≥99,7% (Metall-Basis)
Kupfer(II)oxid

Purity: 99.7% CAS No.: 1317-38-0 Molecular Formula: CuO Formula Weight: 79.55 Physical Form: -200 Mesh Powder Density: 6.3 Refractive Index: 2.63 Appearance: Black Odour: Odourless Melting Point: 1326° Merck Reference: 14,2646 UN No.: 3077 MDL No.: MFCD00010979

Formel: CuO
Siedepunkt: 2000 °C (1013 hPa)
Schmelzpunkt: 1326 °C
Dichte: 6,3
Lagertemperatur: Raumtemperatur
MDL: MFCD00010979
CAS-Nummer: 1317-38-0
EINECS: 215-269-1
UN: 3077
ADR: 9,III
Merck Index: 14,2646

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR