Specpure® Multi Ion Anion IC Standard Solution in H₂O
Lieferant: Thermo Fisher Scientific
41694.AE
41694.AEEA
241
EUR
41694.AE
Specpure® Multi Ion Anion IC Standard Solution in H₂O
Standards
Anionische Standards
Anionische Multi-Element-Standards
Physical Form: Liquid
MDL No.: MFCD00144813
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...