N,N,N',N',N''-Pentamethyldiethylentriamin 98+%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: N1-(2-(Dimethylamino)ethyl)-N1,N2,N2-trimethylethane-1,2-diamine, 1,1,4,7,7-Pentamethyldiethylenetriamine

416950250 416951000 416955000 416950000
ACRO416950250EA 37.4 EUR
ACRO416950250 ACRO416951000 ACRO416955000 416950000.
N,N,N',N',N''-Pentamethyldiethylentriamin 98+%
N,N,N',N',N''-Pentamethyldiethylentriamin
Formel: C₉H₂₃N₃
Molecular Weight: 173.30 g/mol
Schmelzpunkt: 18 °C
Flammpunkt: 72 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00014876
CAS-Nummer: 3030-47-5
EINECS: 221-201-1
UN: 2922
ADR: 8 II

Bestellen

Spezifikation Testergebnisse

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4415 to 1.4435 (20°C, 589 nm)
GC ≥98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR