Arsen(III)tellurid ≥99,999% (Metall-Basis)
Lieferant: Thermo Scientific
|
Gefahr
|
41708.06
41708.06EA
166
EUR
41708.06
Arsen(III)tellurid ≥99,999% (Metall-Basis)
Arsen(III)tellurid
Purity: 99.999%
CAS No.: 12044-54-1
Molecular Formula: As2Te3
Formula Weight: 532.64
Physical Form: -160 Mesh Powder
Density: 6.50
Melting Point: 381°
UN No.: 1557
MDL No.: MFCD00014167
Formel:
As₂Te₃ Molecular Weight: 532,64 g/mol Schmelzpunkt: 381 °C Dichte: 6,50 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00014167 CAS-Nummer: 12044-54-1 EINECS: 234-955-1 UN: 1557 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...