Silicium(II)oxid ≥99,9% (Metall-Basis)

Lieferant: Thermo Scientific

Synonyme: Silicon monoxide, Silicon oxide

41710.22
41710.22EA 200 EUR
41710.22
Silicium(II)oxid ≥99,9% (Metall-Basis)
Silicium(II)oxid

Purity: 99.9% CAS No.: 10097-28-6 Molecular Formula: SiO Formula Weight: 44.09 Physical Form: 3-6mm Pieces Density: 2.13 Melting Point: 1702° Boiling Point: 1880° Merck Reference: 14,8496 MDL No.: MFCD00151536


For vacuum deposition

Formel: SiO
Molecular Weight: 44,08 g/mol
Siedepunkt: 1880 °C
Schmelzpunkt: >1702 °C
Dichte: 2,13
Lagertemperatur: Raumtemperatur
MDL: MFCD00151536
CAS-Nummer: 10097-28-6
EINECS: 233-232-8
Merck Index: 14,8496

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR