Hexane-1,6-bis(tri-n-butylammonium) dihydroxide 20% (w/w) in wässriger Lösung
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
41725.AD
41725.AK
41725.K2
41725.ADEA
68.7
EUR
41725.AD
41725.AK
41725.K2
Hexane-1,6-bis(tri-n-butylammonium) dihydroxide 20% (w/w) in wässriger Lösung
Hexane-1,6-bis(tri-n-butylammonium) dihydroxide
CAS No.: 69762-88-5
Molecular Formula: C30H68N2O2
Formula Weight: 488.87
Physical Form: Liquid
UN No.: UN3267
MDL No.: MFCD00216633
Formel:
C₃₀H₆₈N₂O₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00216633 CAS-Nummer: 69762-88-5 UN: 3267 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...