Specpure® Interference Check Standard AB(ICS-AB) Solution
Lieferant: Thermo Fisher Scientific
41730.AE
41730.AEEA
286
EUR
41730.AE
Specpure® Interference Check Standard AB(ICS-AB) Solution
Standards
Kationischer Standards
Mehrelement-Standards
UN No.: UN3264
MDL No.: MFCD00147272
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...