di-Natriumoxalat ≥99,5% ACS
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Purity: 99.5+%
CAS No.: 62-76-0
Molecular Formula: Na2C2O4
Formula Weight: 134.00
Physical Form: Powder
Density: 2.340
Appearance: White
Odour: Odourless
Melting Point: 250-270° dec.
Solubility: Soluble in water. Insoluble in alcohol
Beilstein Reference: 3631622
Merck Reference: 14,8650
MDL No.: MFCD00012465
Formel:
Na₂C₂O₄ Dichte: 2,340 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00012465 CAS-Nummer: 62-76-0 EINECS: 200-550-3 Merck Index: 14,8650 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...