Graphit, kollodial, Sprühdose Schmälzmittel

Lieferant: Thermo Fisher Scientific
Gefahr

41775.1X
41775.1XEA 57.8 EUR
41775.1X
Graphit, kollodial, Sprühdose Schmälzmittel
Graphite, colloidal

Applications: Dry film lubricant
Air dries rapidly at room temperature and adheres well to most substrates with minimum surface preparation


Appearance: Black UN No.: UN1950 MDL No.: MFCD00144065


Applications: Dry film lubricant

Lagertemperatur: Raumtemperatur
MDL: MFCD00144065
UN: 1950
ADR: 2.1,

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR