Natriumtellurit (IV) ≥99,5% (Metall-Basis)

Lieferant: Thermo Fisher Scientific
Gefahr

41778.09 41778.18
41778.09EA 72.6 EUR
41778.09 41778.18
Natriumtellurit (IV) ≥99,5% (Metall-Basis)
Sodium tellurite(IV)

Purity: 99.5% CAS No.: 10102-20-2 Molecular Formula: Na2O3Te Formula Weight: 221.58 Physical Form: -100 Mesh Powder Appearance: White Solubility: Soluble in water Merck Reference: 14,8685 UN No.: 3284 MDL No.: MFCD00036141

Formel: Na₂O₃Te
Schmelzpunkt: 710 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00036141
CAS-Nummer: 10102-20-2
EINECS: 233-268-4
UN: 3284
ADR: 6.1,II
Merck Index: 14,8685

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR