di-Bortrioxid ≥99,999% (Metall-Basis)

Lieferant: Thermo Fisher Scientific
Gefahr

41786.04 41786.09 41786.18
41786.04EA 40.4 EUR
41786.04 41786.09 41786.18
di-Bortrioxid ≥99,999% (Metall-Basis)
di-Bortrioxid

Purity: 99.999% CAS No.: 1303-86-2 Molecular Formula: B2O3 Formula Weight: 69.62 Physical Form: Powder Density: 2.46 Appearance: White Odour: Odourless Melting Point: ca 450° Boiling Point: 1860° Merck Reference: 14,1337 MDL No.: MFCD00011315

Formel: B₂O₃
Siedepunkt: 1860 °C
Schmelzpunkt: ∼450 °C
Dichte: 2,46
Lagertemperatur: Raumtemperatur
MDL: MFCD00011315
CAS-Nummer: 1303-86-2
EINECS: 215-125-8
REACH: 01-2119486655-24
Merck Index: 14,1337

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR