Wolfram(V)ethanolat
Lieferant: Thermo Fisher Scientific
|
Achtung
|
CAS No.: 26143-11-3
Molecular Formula: C10H25O5W
Formula Weight: 409.15
Flash Point:
Intermediate for sol-gel coatings
Formel:
C₁₀H₂₅O₅W Siedepunkt: 110…115 °C (1 mmHg) Flammpunkt: >110 °C (230 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00078041 CAS-Nummer: 26143-11-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...