Wolfram(VI)ethanolat
Lieferant: Thermo Fisher Scientific
|
Achtung
|
CAS No.: 62571-53-3
Molecular Formula: C12H30O6W
Formula Weight: 454.21
Physical Form: Powder/chunks
Sensitivity: Moisture Sensitive
Appearance: Off-white to brown
Melting Point: 150°
Solubility: Soluble in ethyl acetate
UN No.: 3181
MDL No.: MFCD00156526
Intermediate for sol-gel coatings
Formel:
C₁₀H₂₅O₅W Schmelzpunkt: 150 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00156526 CAS-Nummer: 62571-53-3 UN: 3181 ADR: 4.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...