Chrom ≥99,2% (Metall-Basis), Pulver APS <10 µm

Lieferant: Thermo Fisher Scientific
Achtung

41797.18 41797.30 41797.A1
41797.18EA 76.8 EUR
41797.18 41797.30 41797.A1
Chrom ≥99,2% (Metall-Basis), Pulver APS <10 µm
Chromium

Purity: 99.2% CAS No.: 7440-47-3 Odour: Odourless UN No.: 3089 MDL No.: MFCD00010944

Formel: Cr
Lagertemperatur: Raumtemperatur
MDL: MFCD00010944
CAS-Nummer: 7440-47-3
EINECS: 231-157-5
UN: 3089
ADR: 4.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR