Poly(styrene-divinylbenzene), 1% quervernetzt mit Divinylbenzol 200-400 mesh

Lieferant: Thermo Fisher Scientific

Synonyme: SDVB resin, Styrene-divinylbenzene resin, Styrene-divinylbenzene copolymer, Poly(styrene-co-divinylbenzene), Polystyrene crosslinked with divinylbenzene

41800.14 41800.22
41800.14EA 115 EUR
41800.14 41800.22
Poly(styrene-divinylbenzene), 1% quervernetzt mit Divinylbenzol 200-400 mesh
Poly(styrene-divinylbenzene)

CAS No.: 9003-70-7 Physical Form: Beads Odour: Odourless MDL No.: MFCD00146442


Applications: For the preparation of functionalized beads used in solid-phase synthesis

Formel: [-CH₂CH(C₆H₅)-]x[-CH₂CH[C₆H₄(CHCH₂)]-]y
Molecular Weight: 234.342 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00146442
CAS-Nummer: 9003-70-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR