Platin-Iridium-Legierung (80:20 gew.%) ≥99,9% (Metall-Basis), Folie, Dicke 0.025 mm (0.001 in)
Lieferant: Thermo Fisher Scientific
41805.FF
41805.FI
41805.GH
41805.FFEA
189
EUR
41805.FF
41805.FI
41805.GH
Platin-Iridium-Legierung (80:20 gew.%) ≥99,9% (Metall-Basis), Folie, Dicke 0.025 mm (0.001 in)
Platin-Iridium-Legierung (80:20 gew.%)
Purity: 99.9%
Molecular Formula: Pt:Ir; 80:20 wt%
Physical Form: ≈0.34g/25x25mm
Odour: Odourless
MDL No.: MFCD02091683
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...