Platin-Rhodium-Legierung (90:10 gew.%) ≥99,9% (Metall-Basis), Folie, Dicke 0.1 mm (0.004 in)
Lieferant: Thermo Fisher Scientific
41810.FF
41810.FI
41810.GH
41810.FFEA
2784
EUR
41810.FF
41810.FI
41810.GH
Platin-Rhodium-Legierung (90:10 gew.%) ≥99,9% (Metall-Basis), Folie, Dicke 0.1 mm (0.004 in)
Platin-Rhodium-Legierung (90:10 gew.%)
Purity: 99.9%
Molecular Formula: Pt:Rh; 90:10 wt%
Physical Form: ≈1.25g/25x25mm
Odour: Odourless
MDL No.: MFCD00198186
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...