Platin ≥99,9% (Metall-Basis), Drahtgeflecht 45 mesh, Ø 0.198 mm (0.0078 in)
Lieferant: Thermo Fisher Scientific
Synonyme:
Platinium
41814.FF
41814.FI
41814.GH
41814.FFEA
436
EUR
41814.FF
41814.FI
41814.GH
Platin ≥99,9% (Metall-Basis), Drahtgeflecht 45 mesh, Ø 0.198 mm (0.0078 in)
Platinum
Purity: 99.9%
CAS No.: 7440-06-4
Physical Form: ≈1.61g/25x25mm, Wire Cloth
Odour: Odourless
MDL No.: MFCD00011179
Formel:
Pt Molecular Weight: 195.08 g/mol Siedepunkt: 3827 Schmelzpunkt: 1769 Dichte: 21.447 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011179 CAS-Nummer: 7440-06-4 EINECS: 231-116-1 Merck Index: 12,07689 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...