Kaliumbis(trimethylsilyl)amid 15% (w/w) 0.7 M in Toluol, AcroSeal®
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
418231000
418238000
418231000.EA
109
EUR
418231000.
418238000.
Kaliumbis(trimethylsilyl)amid 15% (w/w) 0.7 M in Toluol, AcroSeal®
Kaliumbis(trimethylsilyl)amid
Formel:
C₆H₁₈KNSi₂ Flammpunkt: 7 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00010330 CAS-Nummer: 40949-94-8 EINECS: 424-100-2 UN: 2920 ADR: 8 I |
Spezifikation Testergebnisse
Appearance | Clear to slightly hazy colorless to amber Solution |
Titration with HClO4 | 0.6 to 0.9 M |
Melting point | 83 to 86 °C |
GC | ≥95 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...