Dicertrisulfid ≥99,9% (Metall-Basis)

Lieferant: Thermo Fisher Scientific
Achtung

41856.06 41856.14
41856.06EA 99.2 EUR
41856.06 41856.14
Dicertrisulfid ≥99,9% (Metall-Basis)
Dicertrisulfid

Purity: 99.9% CAS No.: 12014-93-6 Molecular Formula: Ce2S3 Formula Weight: 376.43 Physical Form: -325 Mesh Powder Density: 5.02 Melting Point: 2100° UN No.: 3134 MDL No.: MFCD00016007

Formel: Ce₂S₃
Schmelzpunkt: 2100 °C
Dichte: 5,02
Lagertemperatur: Raumtemperatur
MDL: MFCD00016007
CAS-Nummer: 12014-93-6
EINECS: 234-603-7
UN: 3134
ADR: 4.3,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR