Bor ≥99,4% (Metall-Basis), kristalline Stücke 1-30 mm (0.039-1.2 in)
Lieferant: Thermo Fisher Scientific
|
Achtung
|
41858.09
41858.18
41858.09EA
260
EUR
41858.09
41858.18
Bor ≥99,4% (Metall-Basis), kristalline Stücke 1-30 mm (0.039-1.2 in)
Boron
Purity: 99.4%
CAS No.: 7440-42-8
Odour: Odourless
MDL No.: MFCD00134034
Formel:
B Dichte: 1,73 g/cm³ (15 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00134034 CAS-Nummer: 7440-42-8 EINECS: 231-151-2 Merck Index: 13,01333 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...