Supelco® Organic Reference Standard, Dimethyl Sulfone
Lieferant: Merck
41867-1G
41867-1GEA 131 EUR
41867-1G
Supelco® Organic Reference Standard, Dimethyl Sulfone
Standards Organische Standards
Standards: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI e.g. NIST or NMIJ. Certified content by quantitative NMR including uncertainty and expiry date are given on the certificate.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=daca034a8ef1b15b0ce6cfd9a844726e0496f0cb&vid=daca034a8ef1b15b0ce6cfd9a844726e0496f0cb&prid=web&clid=SAPDAM