Hafnium(IV)-tert-butanolat ≥99,9% (Metall-Basis ausgeschlossen Zr) (max. 0,5% Zr)

Lieferant: Thermo Scientific
Achtung

Synonyme: Hafnium(IV) t-butoxide, Hafnium(IV)-tert-butanolate

41908.04 41908.18
41908.04EA 229 EUR
41908.04 41908.18
Hafnium(IV)-tert-butanolat ≥99,9% (Metall-Basis ausgeschlossen Zr) (max. 0,5% Zr)
Hafnium(IV)-tert-butanolat

Purity: 99.9% CAS No.: 2172-02-3 Molecular Formula: C16H36HfO4 Formula Weight: 470.94 Flash Point: 28°(82°F) Physical Form: Liquid Density: 1.15 Sensitivity: Air and Moisture Sensitive Appearance: Pale yellow Melting Point: 2° Boiling Point: 90°/5mm Solubility: Soluble in hydrocarbons, reacts with alcohols, ketones, and esters UN No.: 1993 MDL No.: MFCD00070458

Formel: C₁₆H₃₆HfO₄
Molecular Weight: 470,94 g/mol
Siedepunkt: 90 °C (5 mmHg)
Schmelzpunkt: 2 °C
Dichte: 1,15
Flammpunkt: 28 °C (82 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00070458
CAS-Nummer: 2172-02-3
UN: 1993
ADR: 3,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR