Tetrakis(2,2,6,6-tetramethyl-3,5-heptandionato-O,O')hafnium(IV) ≥99,7% (Metall-Basis)
Lieferant: Thermo Fisher Scientific
41909.06
41909.14
41909.06EA
674
EUR
41909.06
41909.14
Tetrakis(2,2,6,6-tetramethyl-3,5-heptandionato-O,O')hafnium(IV) ≥99,7% (Metall-Basis)
Tetrakis(2,2,6,6-tetramethyl-3,5-heptandionato-O,O')hafnium(IV)
Purity: 99.7%
CAS No.: 63370-90-1
Molecular Formula: C44H76HfO8
Formula Weight: 910.5
Physical Form: Powder
Melting Point: 315°
MDL No.: MFCD01863471
Formel:
C₄₄H₇₆HfO₈ Schmelzpunkt: 315 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01863471 CAS-Nummer: 63370-90-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...