Poly(styrene) 10% (w/w) Dispersion in Wasser, Latex-Mikrosphären 1 micron

Lieferant: Thermo Fisher Scientific

41917.7C
41917.7CEA 688 EUR
41917.7C
Poly(styrene) 10% (w/w) Dispersion in Wasser, Latex-Mikrosphären 1 micron
Poly(styrene)

CAS No.: 9003-53-6 Molecular Formula: [-CH2CH(C6H5)-]n Physical Form: Dispersion Appearance: White Odour: Odourless MDL No.: MFCD00084450

Formel: C₈H₈
Lagertemperatur: Raumtemperatur
MDL: MFCD00084450
CAS-Nummer: 9003-53-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR