Zinn(II)chlorid, wasserfrei ≥99%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Purity: 99% min
CAS No.: 7772-99-8
Molecular Formula: SnCl2
Formula Weight: 189.62
Physical Form: Crystalline
Density: 3.95
Melting Point: 247°
Boiling Point: 652°
Solubility: Soluble in water, alkalies, alcohol, MEK, methyl acetate, ether, and acetone
Merck Reference: 14,8783
UN No.: 3260
MDL No.: MFCD00011241
As a reducing agent, in plating reactions, as a galvanizing reagent, an antisludging agent, a stabilizer, and as a catalyst
Formel:
SnCl₂ Siedepunkt: 652 °C Schmelzpunkt: 247 °C Dichte: 3,95 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011241 CAS-Nummer: 7772-99-8 EINECS: 231-868-0 UN: 3260 ADR: 8,III Merck Index: 14,8783 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...