Indium(III)chlorid, wasserfrei ≥99,99% (Metall-Basis)
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Purity: 99.99%
CAS No.: 10025-82-8
Molecular Formula: InCl3
Formula Weight: 221.18
Physical Form: Crystalline
Density: 3.460
Appearance: White
Odour: Odourless
Melting Point: 586°
Boiling Point: 800°
Solubility: Soluble in water
Merck Reference: 14,4958
UN No.: 3260
MDL No.: MFCD00011058
Formel:
Cl₃In Siedepunkt: 800 °C Schmelzpunkt: 586 °C Dichte: 3,460 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011058 CAS-Nummer: 10025-82-8 EINECS: 233-043-0 UN: 3260 ADR: 8,II Merck Index: 14,4958 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...