2-Aminoacetonitrile hydrochloride ≥98%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: Aminoacetonitrile hydrochloride

41984.14
41984.14EA 114 EUR
41984.14
2-Aminoacetonitrile hydrochloride ≥98%
2-Aminoacetonitrile hydrochloride

Purity: 98+% CAS No.: 6011-14-9 Molecular Formula: H2NCH2CN.HCl Formula Weight: 92.53 Physical Form: Powder Sensitivity: Hygroscopic Odour: Odourless Melting Point: 166-171° dec. UN No.: UN3439 MDL No.: MFCD00012850

Formel: C₂H₅ClN₂
Molecular Weight: 92,53 g/mol
Schmelzpunkt: 166…171 °C (Zersetzung)
Lagertemperatur: Raumtemperatur
MDL: MFCD00012850
CAS-Nummer: 6011-14-9
EINECS: 227-865-9
ADR: 6.1,III
Merck Index: 14,412

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR