Magnesiumsulfat Heptahydrat 99-100,5%, GenAR® USP zur parentalen Verwendung, Macron Fine Chemicals™

Lieferant: Avantor Performance Materials


Veränderungsmanagement-Kategorie= R

4200-07EA 0 EUR
4200-07 4200-20
Magnesiumsulfat Heptahydrat 99-100,5%, GenAR® USP zur parentalen Verwendung, Macron Fine Chemicals™
Magnesiumsulfat Heptahydrat
Formel: MgSO₄·7H₂O
MDL: MFCD00149785
CAS-Nummer: 10034-99-8

Bestellen

Spezifikation Testergebnisse

Food GMP Manufactured Product
Meets U.S.P Requirements
GMP Manufactured Product
CAUTION: For Manufacturing, processing or repackaging
Bulk Food Chemical
Bulk Pharmaceutical Chemical
USP - Assay (as MgSO₄) 99.0 - 100.5 %
USP - Identification A Passes Test
USP - Identification B Passes Test
USP - pH (1 in 20) 5.0 - 8.8
USP - Loss on Ignition 40.0 - 52.0 %
USP - Chloride (Cl) ≤ 0.014 %
USP - Selenium (Se) ≤ 0.003 %
USP - Iron (Fe)(µg/g) ≤ 0.5
Solubility (5 in 50) Passes Test
No Class 1,2,3 or other solvents are used or produced in the manufacturing or purification of the product. 99.5 - 100.5 %
Metallic Residues: No metal catalysts or metal reagents, as defined by EMA Guideline EMEA/CHMP/SWP/4446/2000 , are used in the production of this material. Passes Test
FCC - Identification B Passes Test
FCC - Loss on Ignition 40.0 - 52.0 %
FCC - Lead (Pb) = 4 mg/kg
FCC - Selenium (Se) = 0.003 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR