5-Hydroxy-1,4-naphthochinon (Juglon), Calbiochem®

Lieferant: Merck Millipore Calbiochem

420120-250
420120-250EA 84.7 EUR
420120-250
5-Hydroxy-1,4-naphthochinon (Juglon), Calbiochem®
5-Hydroxy-1,4-naphthochinon (Juglon)
Formel: C₁₀H₆O₃
MDL: MFCD00001684
CAS-Nummer: 481-39-0
EINECS: 207-567-5
UN: 2811
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR