2-Hydroxyacetophenon ≥97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
Phenacylhydroxyid, α-Hydroxyacetophenon, ω-Hydroxyacetophenon
unter Argon abgefüllt
42013.06
42013.06EA
80.5
EUR
42013.06
2-Hydroxyacetophenon ≥97%
2-Hydroxyacetophenon
Purity: 97+%
CAS No.: 582-24-1
Molecular Formula: C8H8O2
Formula Weight: 136.15
Physical Form: Powder, Packaged under argon
Sensitivity: Air and Moisture Sensitive
MDL No.: MFCD00041829
Formel:
C₈H₈O₂ Molecular Weight: 136,15 g/mol Siedepunkt: 118…120 °C (11 mmHg) Schmelzpunkt: 86…88 °C Dichte: 1,0963 g/cm³ (99 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00041829 CAS-Nummer: 582-24-1 EINECS: 209-480-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...