Stainless steel (Fe:Cr:Ni:Mo; 67.5:17:13:2.5 wt%), Kugeln, Ø 3 mm (0.12 in) Type 316
Lieferant: Thermo Scientific
42020.KM
42020.KMEA
28.7
EUR
42020.KM
Stainless steel (Fe:Cr:Ni:Mo; 67.5:17:13:2.5 wt%), Kugeln, Ø 3 mm (0.12 in) Type 316
Stainless steel
Molecular Formula: Fe:Cr:Ni:Mo; 67.5:17:13:2.5 wt%
MDL No.: MFCD00148499
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...