Harnstoff 99,0-100,5%, BAKER ANALYZED® ACS, J.T.Baker®

Lieferant: Avantor
Achtung

Synonyme: Urea, Aminoameisensäureamid, Carbamid, Carbonyldiamid

4204-07EA 1876.67 EUR
4204-07 4204-05 4204-01 4204-09
Harnstoff 99,0-100,5%, BAKER ANALYZED® ACS, J.T.Baker®
Harnstoff
Formel: NH₂CONH₂
Siedepunkt: 197 °C (1013 hPa)
Schmelzpunkt: 132,7 °C
Dichte: 1,32 g/cm³
Lagertemperatur: Raumtemperatur
MDL: MFCD00008022
CAS-Nummer: 57-13-6
EINECS: 200-315-5
REACH: 01-2119463277-33
Merck Index: 13,09935
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a9682f71b212a449398841e2beace2d7500b4710&vid=d6057a7ec3d606031d338a300b5d5db30b52d861&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Meets ACS Reagent Chemical Requirements 99.0 - 100.5%
For Laboratory,Research,or Manufacturing Use 132 - 135 °C
Assay 99.0 - 100.5 %
Melting Point 132 - 135 °C
Insoluble Matter ≤0.01 %
Residue after Ignition ≤0.010 %
Sulfate (SO₄) ≤0.001 %
Heavy Metals (as Pb) ≤0.001 %
Trace Impurities - Iron (Fe) ≤0.001 %
Chloride (Cl) ≤5 ppm
Meets Reagent Specifications for testing USP/NF monographs


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR