(1,5-Cyclooctadiene)(pyridine)(tricyclohexylphosphine)iridium(I) hexafluorophosphate ≥99%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Purity: 99%
CAS No.: 64536-78-3
Molecular Formula: C31H50F6IrNP2
Formula Weight: 804.89
Physical Form: Crystalline
Sensitivity: Moisture Sensitive
Melting Point: 175° dec.
Solubility: Slightly soluble in acetone, dichloromethane, ethanol, diethyl ether
MDL No.: MFCD00075097
Formel:
C₃₁H₅₀F₆IrNP₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00075097 CAS-Nummer: 64536-78-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...