Harnstoff 98,0-102,0% USP, J.T.Baker®
Lieferant: Avantor
|
Achtung
|
Synonyme:
Urea, Aminoameisensäureamid, Carbamid, Carbonyldiamid
4206-01EA
164.87
EUR
4206-01
4206-05
4206-07
Harnstoff 98,0-102,0% USP, J.T.Baker®
Harnstoff
- GMP manufactured product
Formel:
NH₂CONH₂ Siedepunkt: 197 °C (1013 hPa) Schmelzpunkt: 132,7 °C Dichte: 1,34 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00008022 CAS-Nummer: 57-13-6 EINECS: 200-315-5 REACH: 01-2119463277-33 Merck Index: 13,09935 |
|
Spezifikation Testergebnisse
GMP Manufactured Product | Passes Test |
Meets U.S.P Requirements | Passes Test |
CAUTION: For Manufacturing, processing or repackaging | 98.0 - 102.0% |
Bulk Pharmaceutical Chemical | ≤0.1% |
USP - Identification A | Passes Test |
USP - Identification B | Passes Test |
USP - Assay (NH₂CONH₂) | 98.0 - 102.0 % |
USP - Residue on Ignition | ≤0.1 % |
USP - Alcohol Insoluble Matter | ≤0.04 % |
USP - Organic impurities: Urea related compound A | ≤0.1 % |
USP - Organic impurities: Any individual unspecified impurity | ≤0.1 % |
USP - Organic impurities: Total impurities | ≤2.0 % |
No Class 1,2,3 or other solvents are used or produced in the | |
manufacturing or purification of the product. | |
Manufactured exclusively for Avantor Performance Materials, LLC., by BioSpectra, Inc., Stroudsburg, PA | |
Elemental Impurities (USP 232, EP 5.20) - Information on elemental | |
impurities for this product is available on the associated Product | |
Regulatory Data Sheet and elemental impurity profile report. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...