Harnstoff 98,0-102,0% USP, J.T.Baker®

Lieferant: Avantor
Achtung

Synonyme: Urea, Aminoameisensäureamid, Carbamid, Carbonyldiamid

4206-01EA 164.87 EUR
4206-01 4206-05 4206-07
Harnstoff 98,0-102,0% USP, J.T.Baker®
Harnstoff

  • GMP manufactured product

Formel: NH₂CONH₂
Siedepunkt: 197 °C (1013 hPa)
Schmelzpunkt: 132,7 °C
Dichte: 1,34 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00008022
CAS-Nummer: 57-13-6
EINECS: 200-315-5
REACH: 01-2119463277-33
Merck Index: 13,09935
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a9682f71b212a449398841e2beace2d7500b4710&vid=d6057a7ec3d606031d338a300b5d5db30b52d861&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

GMP Manufactured Product Passes Test
Meets U.S.P Requirements Passes Test
CAUTION: For Manufacturing, processing or repackaging 98.0 - 102.0%
Bulk Pharmaceutical Chemical ≤0.1%
USP - Identification A Passes Test
USP - Identification B Passes Test
USP - Assay (NH₂CONH₂) 98.0 - 102.0 %
USP - Residue on Ignition ≤0.1 %
USP - Alcohol Insoluble Matter ≤0.04 %
USP - Organic impurities: Urea related compound A ≤0.1 %
USP - Organic impurities: Any individual unspecified impurity ≤0.1 %
USP - Organic impurities: Total impurities ≤2.0 %
No Class 1,2,3 or other solvents are used or produced in the
manufacturing or purification of the product.
Manufactured exclusively for Avantor Performance Materials, LLC., by BioSpectra, Inc., Stroudsburg, PA
Elemental Impurities (USP 232, EP 5.20) - Information on elemental
impurities for this product is available on the associated Product
Regulatory Data Sheet and elemental impurity profile report.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR