1,1,1,3,3,3-Hexafluor-2-propanol 99,8 eluent additive, LC-MS, Fluka™
Lieferant: Honeywell Chemicals
|
Gefahr
|
Synonyme:
1,1,1,3,3,3-Hexafluorisopropanol, HFIP, 1,1,1,3,3,3-Hexafluor-2-hydroxypropan
42060-10X1ML
42060-10X1MLEA
164.08
EUR
42060-10X1ML
1,1,1,3,3,3-Hexafluor-2-propanol 99,8 eluent additive, LC-MS, Fluka™
1,1,1,3,3,3-Hexafluor-2-propanol
Formel:
C₃H₂F₆O Molecular Weight: 168.04 g/mol Siedepunkt: 59 °C Schmelzpunkt: –4 °C Dichte: 1.596 g/cm³ (25 °C) Flammpunkt: >100 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011651 CAS-Nummer: 920-66-1 EINECS: 213-059-4 UN: 3265 ADR: 8 II Merck Index: 13,09598 |
Spezifikation Testergebnisse
Assay (GC) | Min. 99.8 % |
Suitability for the LC-MS | complying |
Non-volatile matter | Max. 5 ppm |
Water (Karl Fischer) | Max. 200 ppm |
Absorbance at 240 nm | Max. 0.01 |
Absorbance at 200 nm | Max. 0.3 |
Aluminium (Al) | Max. 0.5 ppm |
Barium (Ba) | Max. 0.1 ppm |
Bismuth (Bi) | Max. 0.1 ppm |
Calcium (Ca) | Max. 0.5 ppm |
Cadmium (Cd) | Max. 0.05 ppm |
Cobalt (Co) | Max. 0.02 ppm |
Chromium (Cr) | Max. 0.02 ppm |
Copper (Cu) | Max. 0.02 ppm |
Iron (Fe) | Max. 0.1 ppm |
Potassium (K) | Max. 0.5 ppm |
Lithium (Li) | Max. 0.1 ppm |
Magnesium (Mg) | Max. 0.1 ppm |
Manganese (Mn) | Max. 0.02 ppm |
Molybdenum (Mo) | Max. 0.1 ppm |
Sodium (Na) | Max. 1 ppm |
Nickel (Ni) | Max. 0.02 ppm |
Lead (Pb) | Max. 0.1 ppm |
Strontium (Sr) | Max. 0.1 ppm |
Zinc (Zn) | Max. 0.1 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...