Harnstoff 98,0-102,0% USP, J.T.Baker®

Lieferant: Avantor
Achtung

Synonyme: Urea, Aminoameisensäureamid, Carbamid, Carbonyldiamid

Veränderungsmanagement-Kategorie= R

4208-09EA 0 EUR
4208-09 4208-F9
Harnstoff 98,0-102,0% USP, J.T.Baker®
Harnstoff

  • GMP manufactured product

Formel: NH₂CONH₂
Siedepunkt: 197 °C (1013 hPa)
Schmelzpunkt: 132,7 °C
Dichte: 1,32 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00008022
CAS-Nummer: 57-13-6
EINECS: 200-315-5
REACH: 01-2119463277-33
Merck Index: 13,09935
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a9682f71b212a449398841e2beace2d7500b4710&vid=d6057a7ec3d606031d338a300b5d5db30b52d861&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Meets U.S.P Requirements 98.0 - 102.0%
GMP Manufactured Product ≤0.04%
CAUTION: For Manufacturing, processing or repackaging Passes Test
Bulk Pharmaceutical Chemical Passes Test
USP - Assay (NH₂CONH₂) 98.0 - 102.0 %
USP - Alcohol Insoluble Matter ≤ 0.04 %
USP - Identification A Passes Test
USP - Identification B Passes Test
USP - Organic impurities: Urea related compound A ≤ 0.1 %
USP - Organic impurities: Any individual unspecified impurity ≤ 0.1 %
USP - Organic impurities: Total impurities ≤ 2.0 %
USP - Residue on Ignition ≤ 0.1 %
Absorbance of a 5M Solution - 260 nm ≤ 0.050
Absorbance of a 5M Solution - 280 nm ≤ 0.030
Biuret ≤ 0.01
Conductivity of 8.5M Solution at 25°C, (µmho/cm) ≤ 30
Cyanate (CNO) None Detected
Endotoxin Concentration (EU/g) ≤ 2.5
Insoluble Matter ≤ 0.01 %
Water (H₂O) ≤ 0.5 %
Chloride (Cl) ≤ 1 ppm
Trace Impurities - Copper (Cu) ≤ 1 ppm
Trace Impurities - Iron (Fe) ≤ 1 ppm
Trace Impurities - Lead (Pb) ≤ 5 ppm
Ultrapure Bioreagent Specifications
No Class 1,2,3 or other solvents are used or produced in the
manufacturing or purification of the product.
Elemental Impurities (USP 232, EP 5.20) - Information on elemental
impurities for this product is available on the associated Product
Regulatory Data Sheet and elemental impurity profile report.
Manufactured exclusively for Avantor Performance Materials, LLC., by BioSpectra, Inc., Stroudsburg, PA


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR