Organic reference standard, neat, Bisphenol A, (CRM), TraceCERT®, Supelco®
Lieferant: Merck
42088-100MG
42088-100MGEA 83.9 EUR
42088-100MG
Organic reference standard, neat, Bisphenol A, (CRM), TraceCERT®, Supelco®
Standards Organische Standards
Certified Reference material suitable for HPLC and GC analysis. Produced and certified in accordance with ISO/IEC 17025 and ISO 17034.