Pentacarbonylrhenium(I)bromid ≥98%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
42097.03
42097.03EA
292
EUR
42097.03
Pentacarbonylrhenium(I)bromid ≥98%
Pentacarbonylrhenium(I)bromid
Purity: 98%
CAS No.: 14220-21-4
Molecular Formula: C5BrO5Re
Formula Weight: 406.16
Physical Form: Crystalline
Sensitivity: Air and Moisture Sensitive
Appearance: White
Melting Point: ca 90° subl.
UN No.: 3466
MDL No.: MFCD00013295
Formel:
C₅BrO₅Re Lagertemperatur: Raumtemperatur |
MDL:
MFCD00013295 CAS-Nummer: 14220-21-4 EINECS: 238-084-8 UN: 3466 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...