Indium(III)isopropoxid ≥99,9% (Metall-Basis)
Lieferant: Thermo Fisher Scientific
|
Achtung
|
unter Argon abgefüllt
Purity: 99.9%
CAS No.: 38218-24-5
Molecular Formula: C9H21InO3
Formula Weight: 292.08
Physical Form: Powder, Packaged under argon
Sensitivity: Moisture Sensitive
Appearance: White
Melting Point: 145° dec.
UN No.: 3181
MDL No.: MFCD00210318
Precursor used in the preparation of indium oxide containing films
Formel:
C₉H₂₁InO₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00210318 CAS-Nummer: 38218-24-5 UN: 3181 ADR: 4.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...