Nα-(2,4-Dinitro-5-fluorophenyl)-D-valinamide ≥98,0%, LiChropur™ zur chiralen Derivatisierung, Supelco®

Lieferant: Merck
Achtung


M-Clarity™ Qualitätsstufe = MQ100

42100-500MG
42100-500MGEA 984 EUR
42100-500MG
Nα-(2,4-Dinitro-5-fluorophenyl)-D-valinamide ≥98,0%, LiChropur™ zur chiralen Derivatisierung, Supelco®
Nα-(2,4-Dinitro-5-fluorophenyl)-D-valinamide

D-FDVA may be used as derivatizing agent for nullifying the possibility of minor compounds to be diastereomer of xenortide A, during HPLC-MS screening.

Formel: C₁₁H₁₃FN₄O₅
Lagertemperatur: Kühlschrank
MDL: MFCD00155643
CAS-Nummer: 210529-62-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a93e5295d4416c3fd9ac93fbccb3cd38b1824cca&vid=ea6b0f3a50e16e24cdc27d99cc0839cdbeaa7877&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR