n-Hexan ≥95%, Ökologische Qualität
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Purity: 95+%
CAS No.: 110-54-3
Molecular Formula: CH3(CH2)4CH3
Formula Weight: 86.18
Flash Point: -23°(-9F)
Physical Form: Liquid
Density: 0.659
Refractive Index: 1.3750
Odour: Weak, characteristic
Melting Point: -95°
Boiling Point: 69°
Beilstein Reference: 1730733
Merck Reference: 14,4694
UN No.: 1208
MDL No.: MFCD00009520
Pesticide residue, environmental and GC analysis
Formel:
H₃C(CH₂)₄CH₃ Siedepunkt: 69 °C Schmelzpunkt: –95 °C Dichte: 0,659 Flammpunkt: –23 °C (–9F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00009520 CAS-Nummer: 110-54-3 EINECS: 203-777-6 UN: 1208 ADR: 3,II Merck Index: 14,4694 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...