Nα-(2,4-Dinitro-5-fluorophenyl)-L-valinamide ≥98,0%, LiChropur™ zur chiralen Derivatisierung, Supelco®

Lieferant: Merck
Achtung

Synonyme: (S)-2-((5-Fluoro-2,4-dinitrophenyl)amino)-3-methylbutanamide, FDNP-Val-NH₂

M-Clarity™ Qualitätsstufe = MQ100

42102-100MG
42102-100MGEA 146 EUR
42102-100MG
Nα-(2,4-Dinitro-5-fluorophenyl)-L-valinamide ≥98,0%, LiChropur™ zur chiralen Derivatisierung, Supelco®
Nα-(2,4-Dinitro-5-fluorophenyl)-L-valinamide

Nα-(2,4-Dinitro-5-fluorophenyl)-L-valinamide [L-FDVA] is a derivatizing chiral agent.


L-FDVA may be used as derivatizing agent for amino acids in the isolation and identification of microcystins.


Verpackung: Bottomless glass bottle. Contents are inside inserted fused cone.

Formel: C₁₁H₁₃FN₄O₅
Molecular Weight: 300,2431 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00155636
CAS-Nummer: 132679-61-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=475d6dd73f09518a1b8b61b4f3d5d98f5e256656&vid=eaebfa511574b55b390e3a0c034f698428b46a89&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR