Chrom(III)chlorid Hexahydrat ≥98%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Purity: 98%
CAS No.: 10060-12-5
Molecular Formula: CrCl3.6H2O
Formula Weight: 266.45 (158.35anhy)
Physical Form: Crystalline
Density: 1.835
Appearance: Dark Green
Odour: Odourless
Melting Point: 83°
Solubility: Slightly soluble in acetone. Soluble in water, alcohol. Insoluble in ether
Merck Reference: 14,2222
UN No.: 3260
MDL No.: MFCD00149660
Formel:
CrCl₃·6H₂O Schmelzpunkt: 83 °C Dichte: 1,835 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00149660 CAS-Nummer: 10060-12-5 EINECS: 233-038-3 UN: 3260 ADR: 8,III Merck Index: 14,2222 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...