Bis(2-butoxyethyl)ether ≥99%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
42150.AE
42150.AP
42150.AEEA
55
EUR
42150.AE
42150.AP
Bis(2-butoxyethyl)ether ≥99%
Bis(2-butoxyethyl)ether
Purity: 99+%
CAS No.: 112-73-2
Molecular Formula: [CH3(CH2)3OCH2CH2]2O
Formula Weight: 218.74
Flash Point: 117°(243°F)
Physical Form: Liquid
Density: 0.885
Refractive Index: 1.4235
Appearance: Colourless
Melting Point: -60°
Boiling Point: 256°
MDL No.: MFCD00009459
Extraction of precious metals
Formel:
C₁₂H₂₆O₃ Siedepunkt: 256 °C Schmelzpunkt: –60 °C Dichte: 0,885 Flammpunkt: 117 °C (243 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00009459 CAS-Nummer: 112-73-2 EINECS: 204-001-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...